Please login to your online account to display your discounted pricing

anti-S1P1/EDG-1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 340-400 of human EDG-1 was used as the immunogen for this antibody.
Antigen S1P1/EDG-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224622SS

 
novus biologicals canada
NBP224622SS
Unlabeled 0.025mg Each for $139.00

NBP224622

 
novus biologicals canada
NBP224622
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 340-400 of human EDG-1 was used as the immunogen for this antibody.
Antigen S1P1/EDG-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, and 90% homologous in mouse and rat.
Gene Accession No. P21453
Gene ID (Entrez) 1901
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL