Please login to your online account to display your discounted pricing

anti-S100A7/Psoriasin, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human S100A7 was used as the immunogen.
Antigen S100A7/Psoriasin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224506SS

 
novus biologicals canada
NBP224506SS
Unlabeled 0.025mg Each for $109.00

NBP224506

 
novus biologicals canada
NBP224506
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human S100A7 was used as the immunogen.
Antigen S100A7/Psoriasin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The aminoacid sequence used as immunogen is 100% homologus in human (isoforms CRA_a, CRA_c, CRA_e) and chimpanzee, 91% homologus in monkey, 83% homologus in cow and 71% homologus in mouse and rat.
Gene Accession No. P31151
Gene ID (Entrez) 6278
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL