Please login to your online account to display your discounted pricing

anti-S100A7/Psoriasin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224506SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224506SS

  • / Each
 

Description & Specifications

Specifications

Antigen S100A7/Psoriasin
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P31151
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human S100A7 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6278
Test Specificity The aminoacid sequence used as immunogen is 100% homologus in human (isoforms CRA_a, CRA_c, CRA_e) and chimpanzee, 91% homologus in monkey, 83% homologus in cow and 71% homologus in mouse and rat.
Target Species Human
Target Species Primate