Please login to your online account to display your discounted pricing

anti-S100A7/Psoriasin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224506

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224506

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Quantity 0.1mg
Immunogen A portion of amino acids 1-50 of human S100A7 was used as the immunogen.
Gene ID (Entrez) 6278
Test Specificity The aminoacid sequence used as immunogen is 100% homologus in human (isoforms CRA_a, CRA_c, CRA_e) and chimpanzee, 91% homologus in monkey, 83% homologus in cow and 71% homologus in mouse and rat.
Gene Accession No. P31151
Primary or Secondary Primary
Conjugate Unlabeled
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Applications Western Blotting
Regulatory Status RUO
Antigen S100A7/Psoriasin