Please login to your online account to display your discounted pricing

anti-S-Tag Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6005291MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6005291MG

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Conjugate Unlabeled
Antigen S-Tag Epitope Tag
Concentration 1.0mg/mL
Regulatory Status RUO
Formulation Tris-citrate/phosphate buffer, pH 7 to 8
Host Species Goat
Immunogen Goats were immunized with S-Tag (KETAAAKFERQHMDS) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Cross Reactivity Non-species specific
Quantity 1mg
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary