Please login to your online account to display your discounted pricing

anti-S-Tag Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600529

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600529

  • / Each
 

Description & Specifications

Specifications

Antigen S-Tag Epitope Tag
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Non-species specific
Format Affinity Purified
Formulation Tris-citrate/phosphate buffer, pH 7 to 8
Host Species Goat
Immunogen Goats were immunized with S-Tag (KETAAAKFERQHMDS) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Target Species Non-species Specific