missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RUVBL1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human Rvb1 was used as the immunogen.
Antigen RUVBL1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227148 Novus Biologicals Canada
NBP227148
0.1mg Each for $257.41
NBP227148SS Novus Biologicals Canada
NBP227148SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
RUVBL1
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat.
8607
0.5mg/mL
A portion of amino acids 1-50 of human Rvb1 was used as the immunogen.
Polyclonal
Primary
Western Blot
Q9Y265
100μg in 200μL PBS containing 0.05% BSA.