Please login to your online account to display your discounted pricing

anti-RUVBL1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227148SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227148SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9Y265
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene ID (Entrez) 8607
Concentration 0.5mg/mL
Antigen RUVBL1
Format Purified
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat.
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human Rvb1 was used as the immunogen.
Cross Reactivity Human
Applications Western Blotting