missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RPS19BP1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human AROS was used as the immunogen.
Antigen RPS19BP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224621 Novus Biologicals Canada
NBP224621
0.1mg Each for $259.02
NBP224621SS Novus Biologicals Canada
NBP224621SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
RPS19BP1
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human.
91582
0.5mg/mL
A portion of amino acids 1-50 of human AROS was used as the immunogen.
Polyclonal
Primary
Western Blot
Q86WX3
100μg in 200μL PBS containing 0.05% BSA