Please login to your online account to display your discounted pricing

anti-ROR gamma/RORC/NR1F3, Polyclonal, Novus Biologicals™

$109.00 - $373.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human ROR gamma was used as the immunogen.
Antigen ROR gamma/RORC/NR1F3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224503SS

 
novus biologicals canada
NBP224503SS
Unlabeled 0.025mg Each for $109.00

NBP224503

 
novus biologicals canada
NBP224503
Unlabeled 0.1mg Each for $373.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human ROR gamma was used as the immunogen.
Antigen ROR gamma/RORC/NR1F3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), horse, zebrafinch, mouse, monkey, opossum, rhesus monkey, pigeon, wild boar, cow, dog, orangutan, chicken, rat and chimpanzee, 83% homologous in zebrafish and 77% homologous in pufferfish.
Gene ID (Entrez) 6097
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL