Please login to your online account to display your discounted pricing

anti-ROR alpha/NR1F1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224519SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224519SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cow, monkey, rat, opossum and zebrafish, 93% in mouse, 86% in chicken and 80% homologous in chimpanzee and xenopus.
Gene Accession No. P35398
Gene ID (Entrez) 6095
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL
Primary or Secondary Primary
Regulatory Status RUO
Quantity 0.025mg
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Applications Western Blotting
Immunogen A portion of amino acids 80-120 of human ROR alpha was used as the immunogen.
Format Affinity Purified
Antigen ROR alpha/NR1F1
Host Species Rabbit