Please login to your online account to display your discounted pricing

anti-ROR alpha/NR1F1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224519

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224519

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cow, monkey, rat, opossum and zebrafish, 93% in mouse, 86% in chicken and 80% homologous in chimpanzee and xenopus.
Concentration 0.5mg/mL
Gene Accession No. P35398
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Regulatory Status RUO
Immunogen A portion of amino acids 80-120 of human ROR alpha was used as the immunogen.
Gene ID (Entrez) 6095
Primary or Secondary Primary
Applications Western Blotting
Quantity 0.1mg
Format Affinity Purified
Host Species Rabbit
Antigen ROR alpha/NR1F1