Please login to your online account to display your discounted pricing

anti-ROR alpha/NR1F1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224493SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224493SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cow, monkey, horse and chimpanzee and 94% homologous in mouse and rat.
Gene ID (Entrez) 6095
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. P35398
Concentration 0.5mg/mL
Formulation 100μg in 200μL PBS containing 0.05% BSA
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Antigen ROR alpha/NR1F1
Regulatory Status RUO
Immunogen A portion of amino acids 220-270 of human ROR alpha was used as the immunogen.
Format Affinity Purified
Applications Western Blotting
Host Species Rabbit
Quantity 0.025mg