Please login to your online account to display your discounted pricing

anti-ROR alpha/NR1F1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224493

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224493

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P35398
Concentration 0.5mg/mL
Gene ID (Entrez) 6095
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cow, monkey, horse and chimpanzee and 94% homologous in mouse and rat.
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Quantity 0.1mg
Immunogen A portion of amino acids 220-270 of human ROR alpha was used as the immunogen.
Regulatory Status RUO
Format Affinity Purified
Antigen ROR alpha/NR1F1
Host Species Rabbit
Applications Western Blotting