Please login to your online account to display your discounted pricing

anti-ROR alpha/NR1F1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 80-120 of human ROR alpha was used as the immunogen.
Antigen ROR alpha/NR1F1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224519SS

 
novus biologicals canada
NBP224519SS
Unlabeled 0.025mg Each for $109.00

NBP224519

 
novus biologicals canada
NBP224519
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 80-120 of human ROR alpha was used as the immunogen.
Antigen ROR alpha/NR1F1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cow, monkey, rat, opossum and zebrafish, 93% in mouse, 86% in chicken and 80% homologous in chimpanzee and xenopus.
Gene Accession No. P35398
Gene ID (Entrez) 6095
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL