missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Ropporin 1-like, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 58-73 (KDRMEMPTATQKTDTG) and 176-191 (ARLDSDVSPLETESYL) of human ASP protein were used as the immunogen.
Antigen Ropporin 1-like
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056418T Novus Biologicals Canada
NB10056418SS
0.025mg Each for $139.00
NB10056418 Novus Biologicals Canada
NB10056418
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
Ropporin 1-like
Purified
RUO
Reacts with Human. Expected from sequence similarity: Bovine, Chimpanzee, canine and mouse.
83853
0.5mg/mL
Amino acids 58-73 (KDRMEMPTATQKTDTG) and 176-191 (ARLDSDVSPLETESYL) of human ASP protein were used as the immunogen.
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin)
NP_060606
PBS containing 0.2% gelatin