Please login to your online account to display your discounted pricing

anti-ROBO4, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human Robo4 (within residues 900-1007). [Swiss-Prot No. Q8WZ75]
Antigen ROBO4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11058780T

 
novus biologicals canada
NB11058780SS
Unlabeled 0.025mL Each for $109.00

NB11058780

 
novus biologicals canada
NB11058780
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human Robo4 (within residues 900-1007). [Swiss-Prot No. Q8WZ75]
Antigen ROBO4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This recognizes the 85 kDa and 107 kDa isoforms of the ROBO4 human protein.
Gene Accession No. Q8WZ75
Gene ID (Entrez) 54538
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL