Please login to your online account to display your discounted pricing

anti-RNF125, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 100-150 of human TRAC-1 was used as the immunogen.
Antigen RNF125
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223660SS

 
novus biologicals canada
NBP223660SS
Unlabeled 0.025mL Each for $109.00

NBP223660

 
novus biologicals canada
NBP223660
Unlabeled 0.1mL Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 100-150 of human TRAC-1 was used as the immunogen.
Antigen RNF125
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey and dog and 93% homologous in mouse and rat.
Gene Accession No. Q96EQ8
Gene ID (Entrez) 54941
Formulation 100μL of crude rabbit sera with 50% glycerol