Please login to your online account to display your discounted pricing

anti-RNF125, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223660SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223660SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q96EQ8
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Conjugate Unlabeled
Formulation 100μL of crude rabbit sera with 50% glycerol
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Primate
Antigen RNF125
Gene ID (Entrez) 54941
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey and dog and 93% homologous in mouse and rat.
Format Antiserum
Immunogen A portion of amino acid 100-150 of human TRAC-1 was used as the immunogen.
Regulatory Status RUO
Host Species Rabbit
Applications Western Blotting