Please login to your online account to display your discounted pricing

anti-RNA Polymerase II/POLR2A (p Thr4), Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a phosphorylated Threonine (position 4) of the CTD heptad in the human RNA polymerase II protein [UniProt P24928]
Antigen RNA Polymerase II/POLR2A (p Thr4)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149546

 
novus biologicals canada
NBP149546
Unlabeled 0.1mL Each for $309.05

NBP149546SS

 
novus biologicals canada
NBP149546SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a phosphorylated Threonine (position 4) of the CTD heptad in the human RNA polymerase II protein [UniProt P24928]
Antigen RNA Polymerase II/POLR2A (p Thr4)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Dot Blot
Applications Western Blot
Test Specificity This antibody is specific for the phosphorylated Threonine found in position 4 of the RNA polymerase II CTD heptad.
Gene Accession No. P24928
Gene ID (Entrez) 5430
Formulation PBS, 30% glycerol
Concentration 1.1mg/mL