Please login to your online account to display your discounted pricing

anti-RISC, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056648SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056648T

  • / Each
 

Description & Specifications

Specifications

Antigen RISC
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS
Gene Accession No. NP_067639.1
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to aa 28-43 of human RISC.
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 59342
Target Species Human
Target Species Murine
Target Species Rat