Please login to your online account to display your discounted pricing

anti-RIG-I, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 200-250 of human RIG-I was used as the immunogen.
Antigen RIG-I
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227419

 
novus biologicals canada
NBP227419
Unlabeled 0.1mg Each for $255.80

NBP227419SS

 
novus biologicals canada
NBP227419SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 200-250 of human RIG-I was used as the immunogen.
Antigen RIG-I
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for the RIG-I antibody is 100% homologous in human and monkey, 78% homologous in mouse and 82% homologous in rat.
Gene Accession No. O95786
Gene ID (Entrez) 23586
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL