Please login to your online account to display your discounted pricing

anti-RIG-I, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227419SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227419SS

  • / Each
 

Description & Specifications

Specifications

Antigen RIG-I
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene Accession No. O95786
Host Species Rabbit
Immunogen A portion of amino acid 200-250 of human RIG-I was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23586
Test Specificity The amino acid sequence used as immunogen for the RIG-I antibody is 100% homologous in human and monkey, 78% homologous in mouse and 82% homologous in rat.
Target Species Human
Target Species Primate