Please login to your online account to display your discounted pricing

anti-RIG-I, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227419

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227419

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen for the RIG-I antibody is 100% homologous in human and monkey, 78% homologous in mouse and 82% homologous in rat.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Concentration 0.5mg/mL
Format Affinity Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Gene Accession No. O95786
Gene ID (Entrez) 23586
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO
Immunogen A portion of amino acid 200-250 of human RIG-I was used as the immunogen.
Antigen RIG-I