Please login to your online account to display your discounted pricing

anti-Rictor, Polyclonal, Novus Biologicals™

$109.00 - $219.00

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1505-1538 of human Rictor protein were used as the immunogen.
Antigen Rictor
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223806

 
novus biologicals canada
NBP223806
Unlabeled 0.1mL Each for $219.00

NBP223806SS

 
novus biologicals canada
NBP223806SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1505-1538 of human Rictor protein were used as the immunogen.
Antigen Rictor
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Peptide ELISA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog,bovine, chicken, chimpand rhesus monkey, and 93% homologous in xenopus.
Gene Accession No. Q6R327
Gene ID (Entrez) 253260
Formulation 100μL of crude rabbit sera with 50% glycerol