Please login to your online account to display your discounted pricing

anti-Rictor, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223806SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223806SS

  • / Each
 

Description & Specifications

Specifications

Antigen Rictor
Applications Peptide ELISA
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. Q6R327
Host Species Rabbit
Immunogen Within the range of amino acids 1505-1538 of human Rictor protein were used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 253260
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog,bovine, chicken, chimpand rhesus monkey, and 93% homologous in xenopus.
Target Species Bovine
Target Species Canine
Target Species Human
Target Species Murine
Target Species Rat