Please login to your online account to display your discounted pricing

anti-Rictor, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223806

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223806

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene Accession No. Q6R327
Gene ID (Entrez) 253260
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog,bovine, chicken, chimpand rhesus monkey, and 93% homologous in xenopus.
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Peptide ELISA
Formulation 100μL of crude rabbit sera with 50% glycerol
Immunogen Within the range of amino acids 1505-1538 of human Rictor protein were used as the immunogen.
Quantity 0.1mL
Antigen Rictor
Format Antiserum
Host Species Rabbit
Regulatory Status RUO