missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rictor Mouse anti-Human, Mouse, Rat, Clone: 9F1.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051471
Description
Anti-Rictor Antibody, clone 9F1.2 detects level of Rictor and has been published and validated for use in western blotting and immunoprecipitation.Specifications
Rictor | |
Monoclonal | |
Unconjugated | |
NP_689969.2 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunoprecipitation, Western Blot | |
9F1.2 | |
Purified mouse monoclonal IgG2aκ ascites in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
RICTOR | |
Recombinant protein corresponding to human Rictor. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
For Research Use Only