Please login to your online account to display your discounted pricing

anti-Rhox8, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223671

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223671

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen for the FLII antibody is 100% homologous in human, rhesus monkey, horse, mouse, rat and chimpanzee, 94% homologous in cattle and dog, and 89% homologous in chicken.
Applications Immunohistochemistry (Paraffin)
Applications Peptide ELISA
Regulatory Status RUO
Cross Reactivity Mouse
Formulation 100μL; Rhox-8 antibody sera with 50% glycerol
Gene ID (Entrez) 434768
Quantity 0.1mL
Host Species Rabbit
Format Antiserum
Antigen Rhox8
Immunogen Within the range of amino acids 30-60 of mouse Rhox-8 protein were used as the immunogen for the antibody.