missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Rhox8, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 30-60 of mouse Rhox-8 protein were used as the immunogen for the antibody.
Antigen Rhox8
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP223671SS Novus Biologicals Canada
NBP223671SS
0.025mL Each for $109.00
NBP223671 Novus Biologicals Canada
NBP223671
0.1mL Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
Rhox8
Antisera
RUO
The amino acid sequence used as immunogen for the FLII antibody is 100% homologous in human, rhesus monkey, horse, mouse, rat and chimpanzee, 94% homologous in cattle and dog, and 89% homologous in chicken.
100μL; Rhox-8 antibody sera with 50% glycerol
Within the range of amino acids 30-60 of mouse Rhox-8 protein were used as the immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Peptide ELISA
434768