Please login to your online account to display your discounted pricing

anti-Rhox2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223832SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223832SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and 86% homologous in rat.
Gene Accession No. Q9BQY4
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Formulation 100μL; crude rabbit sera with 50% glycerol
Gene ID (Entrez) 75199
Host Species Rabbit
Cross Reactivity Mouse
Quantity 0.025mL
Regulatory Status RUO
Immunogen Within the range of amino acids 1-38 of mouse Rhox-2 protein were used as the immunogen.
Format Antiserum
Antigen Rhox2