Please login to your online account to display your discounted pricing

anti-Rhox2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223832

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223832

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 75199
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and 86% homologous in rat.
Conjugate Unlabeled
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Mouse
Formulation 100μL; crude rabbit sera with 50% glycerol
Gene Accession No. Q9BQY4
Antigen Rhox2
Host Species Rabbit
Immunogen Within the range of amino acids 1-38 of mouse Rhox-2 protein were used as the immunogen.
Quantity 0.1mL
Format Antiserum