Please login to your online account to display your discounted pricing

anti-Rhox2, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1-38 of mouse Rhox-2 protein were used as the immunogen.
Antigen Rhox2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223832

 
novus biologicals canada
NBP223832
Unlabeled 0.1mL Each for $257.41

NBP223832SS

 
novus biologicals canada
NBP223832SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 1-38 of mouse Rhox-2 protein were used as the immunogen.
Antigen Rhox2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and 86% homologous in rat.
Gene Accession No. Q9BQY4
Gene ID (Entrez) 75199
Formulation 100μL; crude rabbit sera with 50% glycerol