Please login to your online account to display your discounted pricing

anti-Rhox11, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 139-171 of mouse Rhox-11 protein were used as the immunogen.
Antigen Rhox11
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223830

 
novus biologicals canada
NBP223830
Unlabeled 0.1mL Each for $257.41

NBP223830SS

 
novus biologicals canada
NBP223830SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 139-171 of mouse Rhox-11 protein were used as the immunogen.
Antigen Rhox11
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and 80% homologous in pufferfish.
Gene ID (Entrez) 194738
Formulation 100μL of crude rabbit sera with 50% glycerol