Please login to your online account to display your discounted pricing

anti-Rhox10, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 90-118 of mouse Rhox-10 protein were used as the immunogen.
Antigen Rhox10
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223834

 
novus biologicals canada
NBP223834
Unlabeled 0.1mL Each for $257.41

NBP223834SS

 
novus biologicals canada
NBP223834SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 90-118 of mouse Rhox-10 protein were used as the immunogen.
Antigen Rhox10
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and rat and 82% homologous in human, rabbit and dog.
Gene ID (Entrez) 434769
Formulation 100μL of crude rabbit sera with 50% glycerol