missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
RHOD, Mouse, Clone: 1E7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant RHOD.
Supplier: Abnova Corporation H00029984M12
Description
Ras homolog, or Rho, proteins interact with protein kinases and may serve as targets for activated GTPase. They play a critical role in muscle differentiation. The protein encoded by this gene binds GTP and is a member of the small GTPase superfamily. It is involved in endosome dynamics and reorganization of the actin cytoskeleton, and it may coordinate membrane transport with the function of the cytoskeleton. [provided by RefSeq
Sequence: MTAAQAAGEEAPPGVRSVKVVLVGDGGCGKTSLLMVFADGAFPESYTPTVFERYMVNLQVKGKPVHLHIWDTAGQDDYDRLRPLFYPDASVLLLCFDVTSPNSFDNIFNRWYPEVNHFCKKVPIIVVGCKTDLRKDKSLVNKLRRNGLEPVTYHRGQEMARSVGAVAYLECSARLHDNVHAVFQEAAEVALSSRGRNFWRRITQGFCVVTSpecifications
RHOD | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC001338 | |
RHOD | |
RHOD (AAH01338, 1 a.a. ∼ 210 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
1E7 | |
Mouse monoclonal antibody raised against a full-length recombinant RHOD. | |
RHOD | |
ARHD/RHOHP1/RHOM/Rho | |
Mouse | |
Affinity chromatography | |
RUO | |
29984 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |