Please login to your online account to display your discounted pricing

anti-RGS6, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 182-198 and 410-426 of human RGS6. It will cross react with mouse (approx. 472 amino acids) and rat RGS11 (approx. 721 amino acids).
Antigen RGS6
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056659T

 
novus biologicals canada
NB10056659SS
Unlabeled 0.05mL Each for $139.00

NB10056659

 
novus biologicals canada
NB10056659
Unlabeled 0.2mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 182-198 and 410-426 of human RGS6. It will cross react with mouse (approx. 472 amino acids) and rat RGS11 (approx. 721 amino acids).
Antigen RGS6
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity An approx. 54 kDa band is observed.
Gene Accession No. NP_004287.3
Gene ID (Entrez) 9628
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL