Please login to your online account to display your discounted pricing

anti-RGS6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056659

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056659

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity An approx. 54 kDa band is observed.
Formulation PBS containing 0.2% gelatin
Gene ID (Entrez) 9628
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Gene Accession No. NP_004287.3
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Antigen RGS6
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 182-198 and 410-426 of human RGS6. It will cross react with mouse (approx. 472 amino acids) and rat RGS11 (approx. 721 amino acids).
Applications Western Blotting
Format Purified
Quantity 0.2mL