missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RGS5, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 56-80 of human RGS5 were used as immunogen.
Antigen RGS5
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224944SS Novus Biologicals Canada
NBP224944SS
0.025mL Each for $139.00
NBP224944 Novus Biologicals Canada
NBP224944
0.1mL Each for $259.02
Specifications Description & Specifications

Specifications

Rabbit
RGS5
Antisera
RUO
O15539
100μL of crude rabbit sera with 50% glycerol
Within the range of amino acids 56-80 of human RGS5 were used as immunogen.
Polyclonal
Primary
Western Blot
8490