Please login to your online account to display your discounted pricing

anti-RGS5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224944SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224944SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL of crude rabbit sera with 50% glycerol
Applications Western Blotting
Conjugate Unlabeled
Immunogen Within the range of amino acids 56-80 of human RGS5 were used as immunogen.
Format Antiserum
Antigen RGS5
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8490
Gene Accession No. O15539
Cross Reactivity Human
Host Species Rabbit
Quantity 0.025mL
Regulatory Status RUO