Please login to your online account to display your discounted pricing

anti-RGS5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224944

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224944

  • / Each
 

Description & Specifications

Specifications

Antigen RGS5
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. O15539
Host Species Rabbit
Immunogen Within the range of amino acids 56-80 of human RGS5 were used as immunogen.
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8490
Target Species Human