missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RGS2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within amino acid residues 1-50 of human RGS2 was used as immunogen.
Antigen RGS2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056736 Novus Biologicals Canada
NB10056736
0.1mg Each for $255.80
NB10056736T Novus Biologicals Canada
NB10056736SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
RGS2
Purified
RUO
Cross reacts with Human, Mouse, Rat. The amino acid sequence used is 86% homologous in mouse and rat.
5997
0.5mg/mL
A synthetic peptide within amino acid residues 1-50 of human RGS2 was used as immunogen.
Polyclonal
Primary
Western Blot
P41220
PBS, 0.2% gelatin