Please login to your online account to display your discounted pricing

anti-RGS18, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 30-62 of human RGS18 protein were used as the immunogen.
Antigen RGS18
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224417

 
novus biologicals canada
NBP224417
Unlabeled 0.1mL Each for $259.02

NBP224417SS

 
novus biologicals canada
NBP224417SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 30-62 of human RGS18 protein were used as the immunogen.
Antigen RGS18
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. Q9NS28
Gene ID (Entrez) 64407
Formulation 100μL of crude rabbit sera with 50% glycerol