Please login to your online account to display your discounted pricing

anti-RGS18, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Amino acids 200-218 of human RGS18 (Genebank accession no NP_570138). were used as immunogen.
Antigen RGS18
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056734

 
novus biologicals canada
NB10056734
Unlabeled 0.1mg Each for $307.00

NB10056734T

 
novus biologicals canada
NB10056734SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 200-218 of human RGS18 (Genebank accession no NP_570138). were used as immunogen.
Antigen RGS18
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Cross reacts with Human. Predicted to react with Mouse and Rat.
Gene Accession No. Q9NS28
Gene ID (Entrez) 64407
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL