Please login to your online account to display your discounted pricing

anti-RGS18, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056734SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056734T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Gene ID (Entrez) 64407
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Test Specificity Cross reacts with Human. Predicted to react with Mouse and Rat.
Immunogen Amino acids 200-218 of human RGS18 (Genebank accession no NP_570138). were used as immunogen.
Regulatory Status RUO
Gene Accession No. Q9NS28
Format Purified
Concentration 0.5mg/mL
Host Species Rabbit
Antigen RGS18
Quantity 0.025mg
Applications Western Blotting