Please login to your online account to display your discounted pricing

anti-RGS18, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056734

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056734

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 64407
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Test Specificity Cross reacts with Human. Predicted to react with Mouse and Rat.
Immunogen Amino acids 200-218 of human RGS18 (Genebank accession no NP_570138). were used as immunogen.
Primary or Secondary Primary
Concentration 0.5mg/mL
Format Purified
Applications Western Blotting
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Human
Gene Accession No. Q9NS28
Host Species Rabbit
Antigen RGS18