missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RGS13, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 67-107 of human RGS13 protein were used as the immunogen.
Antigen RGS13
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224416 Novus Biologicals Canada
NBP224416
0.1mL Each for $259.02
NBP224416SS Novus Biologicals Canada
NBP224416SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
RGS13
Antisera
RUO
O14921
100μL of crude rabbit sera with 50% glycerol
Within the range of amino acids 67-107 of human RGS13 protein were used as the immunogen.
Polyclonal
Primary
Western Blot
6003