Please login to your online account to display your discounted pricing

anti-RGS13, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 111-139 of human RGS13 protein were used as the immunogen.
Antigen RGS13
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224415

 
novus biologicals canada
NBP224415
Unlabeled 0.1mL Each for $259.02

NBP224415SS

 
novus biologicals canada
NBP224415SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 111-139 of human RGS13 protein were used as the immunogen.
Antigen RGS13
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. O14921
Gene ID (Entrez) 6003
Formulation 100μL of crude rabbit sera with 50% glycerol