Please login to your online account to display your discounted pricing

anti-RGS13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224416SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224416SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O14921
Conjugate Unlabeled
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene ID (Entrez) 6003
Primary or Secondary Primary
Quantity 0.025mL
Format Antiserum
Monoclonal or Polyclonal Polyclonal
Immunogen Within the range of amino acids 67-107 of human RGS13 protein were used as the immunogen.
Cross Reactivity Human
Applications Western Blotting
Regulatory Status RUO
Host Species Rabbit
Antigen RGS13