Please login to your online account to display your discounted pricing

anti-RGS13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224416

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224416

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Formulation 100μL of crude rabbit sera with 50% glycerol
Format Antiserum
Immunogen Within the range of amino acids 67-107 of human RGS13 protein were used as the immunogen.
Cross Reactivity Human
Gene ID (Entrez) 6003
Gene Accession No. O14921
Regulatory Status RUO
Antigen RGS13
Host Species Rabbit
Applications Western Blotting
Quantity 0.1mL